Aluminum silicate

Supplier: Thermo Scientific Chemicals

Synonyms: Kaolinite, Silicic acid (H6Si2O7), Aluminum salt (1:2) dihydrate, Aluminum hydroxide silicate (Al2(OH)4(Si2O5)), Aluminum hydroxide silicate (2:4:1), Dialuminum trioxido-Trioxidosilyloxy-silane dihydrate

14231-36 14231-A3
CAAA14231-36EA 84.37 CAD
CAAA14231-36 CAAA14231-A3
Aluminum silicate
Aluminum silicate

Aluminum silicate is used to prepare aluminum-silicate polymer composite (PASiC), which is an inorganic coagulant used in water treatment. It is used as a catalyst for the pyrolysis of rice husk to crude bio -oil, which upgraded to supercritical ethanol. Further, it is used as filler in paper, rubber and in paints.


CAS No.: 12428-46-5 Molecular Formula: Al2Si2O7.2H2O Formula Weight: 258.16 (222.13anhy) Physical Form: Powder Appearance: White to pale grey Odor: Odorless MDL No.: MFCD00062311

Formula: Al₂Si₂O₇.2H₂O
MW: 258.16 g/mol (222.13 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00062311
CAS Number: 12428-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR