Telluric acid dihydrate ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
14197-09
14197-18
CAAA14197-09EA
91.48
CAD
CAAA14197-09
CAAA14197-18
Telluric acid dihydrate ≥99%
Telluric acid dihydrate
Purity: 99+%
CAS No.: 13451-14-4
Molecular Formula: H2O4Te.2H2O
Formula Weight: 229.64 (193.61anhy)
Physical Form: Crystalline
Density: 3.158
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 136°
Boiling Point: 160° dec.
UN No.: 3284
MDL No.: MFCD00150387
Formula:
H₂O₄Te.2H₂0 Boiling Pt: 160 °C (dec.) Melting Pt: 136 °C Density: 3.158 Storage Temperature: Ambient |
MDL Number:
MFCD00150387 CAS Number: 13451-14-4 EINECS: 232-267-6 UN: 3284 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...