Magnesium aluminum zinc, rods, Length: 90 cm (35 in), Ø 9.53 mm (0.375 in)
Supplier: Thermo Scientific Chemicals
14089-RM
14089-RN
CAAA14089-RMEA
205.4
CAD
CAAA14089-RM
CAAA14089-RN
Magnesium aluminum zinc, rods, Length: 90 cm (35 in), Ø 9.53 mm (0.375 in)
Magnesium aluminum zinc
Fieser: 1,1238 15,352 16,366 17,389 18,385 19,382 20,411 21,462
Molecular Formula: Mg:Al:Zn; 96:3:1 wt%
Physical Form: ≈111.7g/90cm
Odor: Odorless
MDL No.: MFCD01074901
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...