Potassium wolframate ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Potassium tungstate, Potassium tungsten oxide
14031-14
14031-22
CAAA14031-14EA
102.7
CAD
CAAA14031-14
CAAA14031-22
Potassium wolframate ≥99.5% (metals basis)
Potassium wolframate
Soluble in water
Purity: 99.5%
CAS No.: 7790-60-5
Molecular Formula: K2WO4
Formula Weight: 326.04
Physical Form: -60 Mesh Powder
Density: 3.12
Melting Point: 921°
Solubility: Soluble in water
Merck Reference: 14,7694
MDL No.: MFCD00049668
Formula:
K₂O₄W Melting Pt: 921 °C Density: 3.12 Storage Temperature: Ambient |
MDL Number:
MFCD00049668 CAS Number: 7790-60-5 EINECS: 232-215-2 Merck Index: 14,7694 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...