Barium zirconate ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
14028-30
14028-A1
CAAA14028-30EA
84.85
CAD
CAAA14028-30
CAAA14028-A1
Barium zirconate ≥99%
Barium zirconate
Purity: 99%
CAS No.: 12009-21-1
Molecular Formula: BaZrO3
Formula Weight: 276.53
Physical Form: 1-2 Micron Powder
Density: 5.52
Odor: Odorless
Melting Point: 2500°
MDL No.: MFCD00014189
Formula:
BaO₃Zr MW: 276.548 g/mol Melting Pt: 2500 °C Density: 5.52 Storage Temperature: Ambient |
MDL Number:
MFCD00014189 CAS Number: 12009-21-1 EINECS: 234-546-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...