Iron disilicide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ferrosilicon, Iron silicide

14022-06 14022-14
CAAA14022-06EA 52.77 CAD
CAAA14022-06 CAAA14022-14
Iron disilicide ≥99.9% (metals basis)
Iron disilicide

Purity: 99.9% CAS No.: 12022-99-0 Molecular Formula: FeSi2 Formula Weight: 112.02 Physical Form: -20 Mesh Odor: Odorless MDL No.: MFCD00016087

Formula: FeSi₂
Melting Pt: 1220 °C
Storage Temperature: Ambient
MDL Number: MFCD00016087
CAS Number: 12022-99-0
EINECS: 234-671-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR