Trimanganese diphosphide ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Manganese phosphide
14020-04
14020-09
CAAA14020-04EA
52.46
CAD
CAAA14020-04
CAAA14020-09
Trimanganese diphosphide ≥99% (metals basis)
Trimanganese diphosphide
Purity: 99%
CAS No.: 12263-33-1
Molecular Formula: Mn3P2 (mixture of MnP & Mn2P)
Formula Weight: 226.75
Physical Form: -100 Mesh Powder
Odor: Odorless
MDL No.: MFCD00064736
Formula:
Mn₃P₂ MW: 226.75 g/mol Melting Pt: 1095 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064736 CAS Number: 12263-33-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...