Iron silicide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

14019-14 14019-22
CAAA14019-14EA 197.5 CAD
CAAA14019-14 CAAA14019-22
Iron silicide ≥99.9% (metals basis)
Iron silicide

Purity: 99.9% CAS No.: 12022-95-6 Molecular Formula: FeSi Formula Weight: 83.93 Physical Form: -20 Mesh Odor: Odorless MDL No.: MFCD00016086

Formula: FeSi
Melting Pt: 1410 °C
Storage Temperature: Ambient
MDL Number: MFCD00016086
CAS Number: 12022-95-6
EINECS: 234-671-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR