Nickel(II) formate dihydrate

Supplier: Thermo Scientific Chemicals
Danger

13801-22 13801-36 13801-A3
CAAA13801-22EA 74.89 CAD
CAAA13801-22 CAAA13801-36 CAAA13801-A3
Nickel(II) formate dihydrate
Nickel(II) formate dihydrate

Applications: Nickel catalyst preparations
In the range 180-200° decomposes to Ni, CO, CO2, H2, H2O, CH3. Partially soluble in water


CAS No.: 15694-70-9 Molecular Formula: Ni(HCO2)2.2H2O Formula Weight: 184.77 (148.74anhy) Physical Form: Powder Density: 2.15 Melting Point: 130-140° -2H2O Solubility: In the range 180-200° decomposes to Ni, CO, CO2, H2, H2O, CH4. Partially soluble in water UN No.: 3077 MDL No.: MFCD00153034


Applications: Nickel catalyst preparations

Formula: Ni(HCO₂)₂·2H₂O
Melting Pt: 130…140 °C (-2H₂O)
Density: 2.15
Storage Temperature: Ambient
MDL Number: MFCD00153034
CAS Number: 15694-70-9
EINECS: 222-101-0
UN: 3077
ADR: 9,III
Merck Index: 14,6509

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR