Nickel (silver coated) (Ni:Ag; 85:15 wt%) ≥99.8% (metals basis), spheres -250 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
13793-14
13793-22
13793-36
CAAA13793-14EA
99.38
CAD
CAAA13793-14
CAAA13793-22
CAAA13793-36
Nickel (silver coated) (Ni:Ag; 85:15 wt%) ≥99.8% (metals basis), spheres -250 mesh
Nickel (silver coated)
Purity: 99.8%
CAS No.: 7440-02-0
Molecular Formula: Ni:Ag; 85:15 wt%
Odor: Odorless
MDL No.: MFCD02091733
Formula:
Ni Storage Temperature: Ambient |
MDL Number:
MFCD02091733 CAS Number: 7440-02-0 EINECS: 231-111-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...