Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), soluble -325 mesh, Thickness: 0.4-1.2 µm Type 316-L
Supplier: Thermo Scientific Chemicals
13787-30
13787-A1
CAAA13787-30EA
85.79
CAD
CAAA13787-30
CAAA13787-A1
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), soluble -325 mesh, Thickness: 0.4-1.2 µm Type 316-L
Stainless steel
Molecular Formula: Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
MDL No.: MFCD00148499
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...