Neodymium boron iron, magnet alloy additive Nd:B:Fe (15:8:77) atomic%, REacton®
Supplier: Thermo Scientific Chemicals
13642-18
13642-30
CAAA13642-18EA
131.61
CAD
CAAA13642-18
CAAA13642-30
Neodymium boron iron, magnet alloy additive Nd:B:Fe (15:8:77) atomic%, REacton®
Neodymium boron iron, magnet alloy additive
Molecular Formula: Nd:B:Fe; 15:8:77 atomic%; Magnet Alloy Additive
Physical Form: Broken Lump
Odour: Odourless
MDL No.: MFCD02091729
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...