Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 40 mesh woven from 0.25mm (0.01in) Type 304
Supplier: Thermo Scientific Chemicals
13478-CH
13478-RR
13478-RU
CAAA13478-CHEA
149.47
CAD
CAAA13478-CH
CAAA13478-RR
CAAA13478-RU
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 40 mesh woven from 0.25mm (0.01in) Type 304
Stainless steel
Open area: 36%; Width of opening: 0.015in
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Physical Form: Wire Cloth
MDL No.: MFCD00148499
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...