Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 80 mesh woven from 0.127mm (0.005in) Type 304
Supplier: Thermo Scientific Chemicals
13477-CH
13477-RR
13477-RU
CAAA13477-CHEA
184.86
CAD
CAAA13477-CH
CAAA13477-RR
CAAA13477-RU
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 80 mesh woven from 0.127mm (0.005in) Type 304
Stainless steel
Open area: 36%; Width of opening: 0.0075in
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Physical Form: Wire Cloth
MDL No.: MFCD00148499
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...