Stainless steel (Fe:Cr:Ni; 73:18:9 wt%), rods, Ø 3.175 mm (0.125 in) Type 303
Supplier: Thermo Scientific Chemicals
13475-BV
13475-RM
CAAA13475-BVEA
51.67
CAD
CAAA13475-BV
CAAA13475-RM
Stainless steel (Fe:Cr:Ni; 73:18:9 wt%), rods, Ø 3.175 mm (0.125 in) Type 303
Stainless steel
Molecular Formula: Fe:Cr:Ni; 73:18:9 wt%
MDL No.: MFCD00148499
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...