Diamond (synthetic) ≥99.9% (metals basis), powder <1 micron

Supplier: Thermo Scientific Chemicals

13401-03 13401-06
CAAA13401-03EA 119.13 CAD
CAAA13401-03 CAAA13401-06
Diamond (synthetic) ≥99.9% (metals basis), powder <1 micron
Diamond

Applications: Diamond powder is generally used as an abrasive to grind and polish hard surfaces.


Purity: 99.9% MDL No.: MFCD00211867


Applications: Diamond powder is generally used as an abrasive to grind and polish hard surfaces.

Storage Temperature: Ambient
MDL Number: MFCD00211867
CAS Number: 7782-40-3
EINECS: 231-953-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR