Methylmercury hydroxide 1 M in water

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methylmercury(II) hydroxide

13395-AB 13395-AD
CAAA13395-ABEA 331.8 CAD
CAAA13395-AB CAAA13395-AD
Methylmercury hydroxide 1 M in water
Methylmercury hydroxide

Applications: defoliant and epoxidation catalyst, as denaturing agent


CAS No.: 1184-57-2 Molecular Formula: CH4HgO Formula Weight: 232.63 Physical Form: Liquid Appearance: Colorless UN No.: 2024 MDL No.: MFCD00013924


As a defoliant and epoxidation catalyst, as denaturing agent

Formula: CH₄HgO
MW: 232.632 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013924
CAS Number: 1184-57-2
EINECS: 214-667-2
UN: 2024
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR