Dysprosium(III) 2,4-pentanedionate hydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Dysprosium (III) acetylacetonate hydrate

13219-06 13219-14
CAAA13219-06EA 79 CAD
CAAA13219-06 CAAA13219-14
Dysprosium(III) 2,4-pentanedionate hydrate ≥99.9% (REO, rare earth oxide basis)
Dysprosium (III) acetylacetonate hydrate

Purity: 99.9% CAS No.: 14637-88-8 Molecular Formula: C15H21DyO6.xH2O Formula Weight: 459.83(anhy) Physical Form: Powder Sensitivity: Hygroscopic MDL No.: MFCD00013489

Formula: C₁₅H₂₁DyO₆
MW: 459.83 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00013489
CAS Number: 14637-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR