Erbium(III)-2,4-pentanedionate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
Synonyms:
Erbium acetylacetonate
13214-04
13214-09
CAAA13214-04EA
172.22
CAD
CAAA13214-04
CAAA13214-09
Erbium(III)-2,4-pentanedionate ≥99.9% (REO, rare earth oxide basis), REacton®
Erbium(III)-2,4-pentanedionate
Purity: 99.9%
CAS No.: 14553-08-3
Molecular Formula: C15H21ErO6
Formula Weight: 464.59
Physical Form: Powder
Sensitivity: Hygroscopic
Odor: Odorless
MDL No.: MFCD00013490
Formula:
C₁₅H₂₁ErO₆ MW: 464.59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013490 CAS Number: 14553-08-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...