Europium(III) carbonate hydrate ≥99.99%, REacton®

Supplier: Thermo Scientific Chemicals

13212-03 13212-06 13212-14
CAAA13212-03EA 64.15 CAD
CAAA13212-03 CAAA13212-06 CAAA13212-14
Europium(III) carbonate hydrate ≥99.99%, REacton®
Europium(III) carbonate hydrate

Purity: 99.99% CAS No.: 86546-99-8 Molecular Formula: Eu2(CO3)3.xH2O Formula Weight: 483.95(anhy) Physical Form: Powder Odor: Odorless MDL No.: MFCD00149702

Formula: Eu₂(CO₃)₃·xH₂O
MW: 483.95 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149702
CAS Number: 86546-99-8
EINECS: 227-582-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR