Europium(III) acetylacetonate hydrate ≥95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
13209-04
13209-06
13209-09
CAAA13209-04EA
248.06
CAD
CAAA13209-04
CAAA13209-06
CAAA13209-09
Europium(III) acetylacetonate hydrate ≥95%
Europium(III) acetylacetonate hydrate
Purity: 99.9%
CAS No.: 181266-82-0
Molecular Formula: C15H21EuO6.xH2O
Formula Weight: 449.29(anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Melting Point: 140° dec.
MDL No.: MFCD00192170
Formula:
C15H21EuO6•xH2O MW: 449.29 g/mol (anhy) Melting Pt: 140 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00192170 CAS Number: 181266-82-0 EINECS: 238-185-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...