Gadolinium(III) carbonate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

13207-14 13207-22
CAAA13207-14EA 192.76 CAD
CAAA13207-14 CAAA13207-22
Gadolinium(III) carbonate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Gadolinium(III) carbonate hydrate

Purity: 99.99% CAS No.: 38245-36-2 Molecular Formula: Gd2(CO3)3.xH2O Formula Weight: 494.53(anhy) Physical Form: Powder MDL No.: MFCD00150240

Formula: Gd₂(CO₃)₃.xH₂O
MW: 494.53 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150240
CAS Number: 38245-36-2
EINECS: 227-583-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR