Gadolinium(III)-2,4-pentanedionate hydrate ≥99.9%, REacton®

Supplier: Thermo Scientific Chemicals

13204-06 13204-14
CAAA13204-06EA 105.86 CAD
CAAA13204-06 CAAA13204-14
Gadolinium(III)-2,4-pentanedionate hydrate ≥99.9%, REacton®
Gadolinium(III)-2,4-pentanedionate hydrate

Purity: 99.9% CAS No.: 64438-54-6 Molecular Formula: C15H21GdO6.xH2O Formula Weight: 454.58(anhy) Physical Form: Powder Sensitivity: Hygroscopic MDL No.: MFCD00013493

Formula: C₁₅H₂₁GdO₆.xH₂O
MW: 454.58 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00013493
CAS Number: 64438-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR