Holmium(III)-2,4-pentanedionate ≥99.9%, REacton®
Supplier: Thermo Scientific Chemicals
13199-04
13199-09
CAAA13199-04EA
167.48
CAD
CAAA13199-04
CAAA13199-09
Holmium(III)-2,4-pentanedionate ≥99.9%, REacton®
Holmium(III)-2,4-pentanedionate
Purity: 99.9%
CAS No.: 14589-33-4
Molecular Formula: C15H21HoO6
Formula Weight: 462.26
Physical Form: Powder
Sensitivity: Hygroscopic
Odor: Odorless
MDL No.: MFCD00050176
Formula:
C₁₅H₂₁HoO₆ MW: 462.26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00050176 CAS Number: 14589-33-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...