Lanthanum(III) bromide hydrate ≥99.99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
13198-18
13198-30
CAAA13198-18EA
133.04
CAD
CAAA13198-18
CAAA13198-30
Lanthanum(III) bromide hydrate ≥99.99%
Lanthanum(III) bromide hydrate
Purity: 99.99%
CAS No.: 224183-16-8
Molecular Formula: LaBr3.xH2O
Formula Weight: 378.62(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
MDL No.: MFCD00798151
Formula:
Br₃H₂LaO MW: 396.632 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00798151 CAS Number: 224183-16-8 EINECS: 236-896-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...