Lanthanum(III) carbonate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

13197-18 13197-30
CAAA13197-18EA 150.1 CAD
CAAA13197-18 CAAA13197-30
Lanthanum(III) carbonate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Lanthanum (III) carbonate hydrate

Purity: 99.99% CAS No.: 54451-24-0 Molecular Formula: La2(CO3)3.xH2O Formula Weight: 457.85(anhy) Physical Form: Powder/Lump Density: 2.6-2.7 Merck Reference: 14,5364 MDL No.: MFCD00011069

Formula: La₂(CO₃)₃.xH₂O
MW: 457.85 g/mol (anhy)
Density: 2.6…2.7
Storage Temperature: Ambient
MDL Number: MFCD00011069
CAS Number: 54451-24-0
EINECS: 209-599-5
Merck Index: 14,5364

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR