Lanthanum(III) 2,4-pentanedionate hydrate ≥99.9%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Lanthanum(III) acetylacetonate hydrate, Tris(2,4-pentanedionato)lanthanum(III) hydrate, Acetylacetone Lanthanum(III) hydrate
13194-14
13194-22
CAAA13194-14EA
150.57
CAD
CAAA13194-14
CAAA13194-22
Lanthanum(III) 2,4-pentanedionate hydrate ≥99.9%
Lanthanum(III) 2,4-pentanedionate hydrate
Purity: 99.9%
CAS No.: 64424-12-0
Molecular Formula: C15H21LaO6.xH2O
Formula Weight: 436.24(anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White to pale yellow
Melting Point: 143°
MDL No.: MFCD00149057
Formula:
C₁₅H₂₁LaO₆·xH₂O MW: 436.24 g/mol (anhy) Melting Pt: 143 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149057 CAS Number: 64424-12-0 EINECS: 238-187-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...