Neodymium(III)-2,4-pentanedionate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Neodymium(III) acetylacetonate

13184-04 13184-09
CAAA13184-04EA 86.43 CAD
CAAA13184-04 CAAA13184-09
Neodymium(III)-2,4-pentanedionate ≥99.9% (REO, rare earth oxide basis)
Neodymium(III)-2,4-pentanedionate

Purity: 99.9% CAS No.: 14589-38-9 Molecular Formula: C15H21NdO6 Formula Weight: 441.57 Physical Form: Powder Odor: Odorless Melting Point: 150-152° MDL No.: MFCD00013498

Formula: C₁₅H₂₁NdO₆
MW: 441.57 g/mol
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00013498
CAS Number: 14589-38-9
EINECS: 238-631-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR