Samarium(III) carbonate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
13177-14
13177-22
CAAA13177-06EA
51.07
CAD
CAAA13177-06
CAAA13177-14
CAAA13177-22
Samarium(III) carbonate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Samarium(III) carbonate hydrate
Purity: 99.99%
CAS No.: 38245-37-3
Molecular Formula: Sm2(CO3)3.xH2O
Formula Weight: 480.73(anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Melting Point:
Formula:
Sm₂(CO₃)₃.xH₂O MW: 480.73 g/mol (anhy) Melting Pt: >500 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00150444 CAS Number: 38245-37-3 EINECS: 227-581-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...