Yttrium(III) carbonate hydrate ≥99.99%, REacton®

Supplier: Thermo Scientific Chemicals

13157-14 13157-22
CAAA13157-14EA 222.78 CAD
CAAA13157-14 CAAA13157-22
Yttrium(III) carbonate hydrate ≥99.99%, REacton®
Yttrium(III) carbonate hydrate

Purity: 99.99% CAS No.: 38245-39-5 Molecular Formula: Y2(CO3)3.xH2O (x≈3) Formula Weight: 357.84(anhy) Physical Form: Powder MDL No.: MFCD00149943

Formula: Y₂(CO₃)₃·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00149943
CAS Number: 38245-39-5
EINECS: 209-119-4
Merck Index: 14,10107

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR