Iron(II) titanium oxide ≥99.8% (metals basis)

Supplier: Thermo Scientific Chemicals

13137-18 13137-30
CAAA13137-18EA 274.92 CAD
CAAA13137-18 CAAA13137-30
Iron(II) titanium oxide ≥99.8% (metals basis)
Iron(II) titanium oxide

Purity: 99.8+% CAS No.: 12022-71-8 Molecular Formula: FeTiO3 Formula Weight: 151.75 Physical Form: -100 Mesh Powder Odor: Odorless MDL No.: MFCD00064704

Formula: FeTiO₃
Storage Temperature: Ambient
MDL Number: MFCD00064704
CAS Number: 12022-71-8
EINECS: 234-667-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR