Manganese(II) titanium oxide ≥99.9%
Supplier: Thermo Scientific Chemicals
13133-09
13133-18
CAAA13133-09EA
267.02
CAD
CAAA13133-09
CAAA13133-18
Manganese(II) titanium oxide ≥99.9%
Manganese(II) titanium oxide
Purity: 99.9%
CAS No.: 12032-74-5
Molecular Formula: MnTiO3
Formula Weight: 150.84
Physical Form: -100 Mesh Powder
Odor: Odorless
Melting Point: 1360°
MDL No.: MFCD00016216
Formula:
MnTiO₃ MW: 150.84 g/mol Melting Pt: 1360 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016216 CAS Number: 12032-74-5 EINECS: 234-776-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...