Niobium disilicide ≥99.85% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
13100-14
13100-22
CAAA13100-14EA
237
CAD
CAAA13100-14
CAAA13100-22
Niobium disilicide ≥99.85% (metals basis)
Niobium disilicide
Purity: 99.85%
CAS No.: 12034-80-9
Molecular Formula: NbSi2
Formula Weight: 149.08
Physical Form: -325 Mesh Powder
Density: 5.7
Melting Point: 1950°
MDL No.: MFCD00151537
Formula:
NbSi₂ Melting Pt: 1950 °C Density: 5,7 Storage Temperature: Ambient |
MDL Number:
MFCD00151537 CAS Number: 12034-80-9 EINECS: 234-812-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...