Tantalum nitride ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals

13093-09 13093-18
CAAA13093-09EA 108.07 CAD
CAAA13093-09 CAAA13093-18
Tantalum nitride ≥99.5% (metals basis)
Tantalum nitride

Purity: 99.5% CAS No.: 12033-62-4 Molecular Formula: TaN Formula Weight: 194.95 Physical Form: -325 Mesh Powder Odor: Odorless MDL No.: MFCD00049568

Formula: NTa
Melting Pt: 3360 °C
Storage Temperature: Ambient
MDL Number: MFCD00049568
CAS Number: 12033-62-4
EINECS: 234-788-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR