Titanium disilicide ≥99.5% (metals basis), powder
Supplier: Thermo Scientific Chemicals
|
Warning
|
13088-18
13088-30
CAAA13088-18EA
192.76
CAD
CAAA13088-18
CAAA13088-30
Titanium disilicide ≥99.5% (metals basis), powder
Titanium disilicide
Purity: 99.5%
CAS No.: 12039-83-7
Molecular Formula: Si2Ti
Formula Weight: 104.07
Physical Form: -200 Mesh Powder
Appearance: Grey
Melting Point: 1760°
UN No.: 3178
MDL No.: MFCD00151541
Formula:
Si₂Ti Melting Pt: 1760 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151541 CAS Number: 12039-83-7 EINECS: 234-904-3 UN: 3178 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...