Pentatitanium trisilicide ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

13087-18 13087-30
CAAA13087-18EA 219.62 CAD
CAAA13087-18 CAAA13087-30
Pentatitanium trisilicide ≥99.5% (metals basis)
Pentatitanium trisilicide

Purity: 99.5% CAS No.: 12067-57-1 Molecular Formula: Ti5Si3 Formula Weight: 323.76 Physical Form: -325 Mesh Powder Odor: Odorless UN No.: 3178 MDL No.: MFCD01310208

Formula: Si₃Ti₅
Melting Pt: 2130 °C
Storage Temperature: Ambient
MDL Number: MFCD01310208
CAS Number: 12067-57-1
EINECS: 235-082-9
UN: 3178
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR