Dodecacarbonyltriiron ≥96% (dry weight) stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Triiron dodecacarbonyl, Iron dodecacarbonyl

Stabilization: Stabilised with methanol 5-10%

13059-04 13059-09 13059-18
CAAA13059-04EA 53.72 CAD
CAAA13059-04 CAAA13059-09 CAAA13059-18
Dodecacarbonyltriiron ≥96% (dry weight) stabilized
Dodecacarbonyltriiron

Reducing agent (reduces NO2), catalyst for alkyne trimerization.


Purity: 96% CAS No.: 17685-52-8 Molecular Formula: C12Fe3O12 Formula Weight: 503.67 Physical Form: Crystalline Density: 2.00 Appearance: Black Melting Point: 140° dec. UN No.: 3179 MDL No.: MFCD00135617


Reducing agent (reduces NO2), catalyst for alkyne trimerization.

Formula: C₁₂Fe₃O₁₂
MW: 503.66 g/mol
Melting Pt: 140 °C (dec.)
Density: 2.00
Storage Temperature: Ambient
MDL Number: MFCD00135617
CAS Number: 17685-52-8
EINECS: 241-668-5
UN: 3179
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR