Calcium wolframate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Calcium tungstate, Calcium tungsten oxide

13051-14 13051-22 13051-36
CAAA13051-14EA 64.78 CAD
CAAA13051-14 CAAA13051-22 CAAA13051-36
Calcium wolframate 98%
Calcium wolframate

Insoluble in H2O. Decomposes by hot HCl, HNO3


Purity: 98% CAS No.: 7790-75-2 Molecular Formula: CaWO4 Formula Weight: 287.93 Physical Form: -325 Mesh Powder Melting Point: 1620° MDL No.: MFCD00010913

Formula: CaO₄W
MW: 287.92 g/mol
Melting Pt: 1620 °C
Storage Temperature: Ambient
MDL Number: MFCD00010913
CAS Number: 7790-75-2
EINECS: 232-219-4
Merck Index: 14,1712

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR