Bismuth germanium oxide ≥99.9995% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

13029-14 13029-22
CAAA13029-14EA 263.86 CAD
CAAA13029-14 CAAA13029-22
Bismuth germanium oxide ≥99.9995% (metals basis), Puratronic®
Bismuth germanium oxide

Purity: 99.9995% CAS No.: 12233-56-6 Molecular Formula: Bi4Ge3O12 Formula Weight: 1245.68 Physical Form: Powder Appearance: White Odor: Odorless MDL No.: MFCD00064629

Formula: Bi₄Ge₃O₁₂
MW: 1245.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064629
CAS Number: 12233-56-6
EINECS: 235-457-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR