Cobalt(II) oxalate dihydrate ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
13012-09
13012-18
CAAA13012-09EA
140.78
CAD
CAAA13012-09
CAAA13012-18
Cobalt(II) oxalate dihydrate ≥99.995% (metals basis), Puratronic®
Cobalt (II) oxalate dihydrate
Purity: 99.995%
CAS No.: 5965-38-8
Molecular Formula: C2CoO4.2H2O
Formula Weight: 182.98 (146.95anhy)
Physical Form: Powder
Density: 3.021
Appearance: Pink
Odor: Odorless
Melting Point: 229-231° dec.
Merck Reference: 14,2445
UN No.: 3288
MDL No.: MFCD00050793
Formula:
C₂CoO₄·2H₂O Melting Pt: 229…231 °C (dec.) Density: 3.021 Storage Temperature: Ambient |
MDL Number:
MFCD00050793 CAS Number: 5965-38-8 EINECS: 212-409-3 UN: 3288 ADR: 6.1,III Merck Index: 14,2445 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...