Strontium acetate hydrate ≥99.9965% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
12990-06
12990-14
CAAA12990-06EA
142.36
CAD
CAAA12990-06
CAAA12990-14
Strontium acetate hydrate ≥99.9965% (metals basis), Puratronic®
Strontium acetate hydrate
Converts to SrCO₃ on ignition
Purity: 99.9965%
CAS No.: 543-94-2
Molecular Formula: C4H6O4Sr.xH2O
Formula Weight: 205.71(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Melting Point: 150° -0.5H2O
Beilstein Reference: 3692534
MDL No.: MFCD00036392
Formula:
C₄H₆O₄Sr·xH₂O Melting Pt: 150 °C (-0.5H₂O) Storage Temperature: Ambient |
MDL Number:
MFCD00036392 CAS Number: 543-94-2 EINECS: 208-854-8 Merck Index: 14,8836 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...