Zinc oxalate dihydrate ≥99.999%, Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Zinc(II) ethane-1,2-dioate dihydrate

12985-14 12985-22
CAAA12985-14EA 267.02 CAD
CAAA12985-14 CAAA12985-22
Zinc oxalate dihydrate ≥99.999%, Puratronic®
Zinc oxalate dihydrate

Purity: 99.999% CAS No.: 4255-07-6 Molecular Formula: C2O4Zn.2H2O Formula Weight: 189.42 (153.39anhy) Physical Form: Powder Density: 3.28 Appearance: White UN No.: 3288 MDL No.: MFCD00150611

Formula: ZnC₂O₄.2H₂O
MW: 189.42 g/mol (153.39 anhy)
Density: 3.28
Storage Temperature: Ambient
MDL Number: MFCD00150611
CAS Number: 4255-07-6
EINECS: 208-934-2
UN: 3288
ADR: 6.1 III
Merck Index: 14,10146

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR