Ytterbium(III) nitrate hydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ytterbium trinitrate hydrate

12901-09 12901-18
CAAA12901-09EA 86.27 CAD
CAAA12901-09 CAAA12901-18
Ytterbium(III) nitrate hydrate ≥99.9% (REO, rare earth oxide basis)
Ytterbium(III) nitrate hydrate

Purity: 99.9% Molecular Formula: Yb(NO3)3.xH2O Formula Weight: 359.05(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White UN No.: 1477 MDL No.: MFCD00149883

Formula: Yb(NO₃)₃.xH₂O
MW: 359.05 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149883
EINECS: 237-384-6
UN: 1477
ADR: 5.1,III
Merck Index: 14,10105

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR