Manganese(II) sulfide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
12835-06
12835-14
12835-22
CAAA12835-06EA
64.78
CAD
CAAA12835-06
CAAA12835-14
CAAA12835-22
Manganese(II) sulfide ≥99.9% (metals basis)
Manganese (II) sulphide
Purity: 99.9%
CAS No.: 18820-29-6
Molecular Formula: MnS
Formula Weight: 87.00
Physical Form: -325 Mesh Powder
Density: 3.99
Appearance: Green to pink
Melting Point: 1610° dec.
Merck Reference: 14,5740
MDL No.: MFCD00014210
Formula:
MnS MW: 87.00 g/mol Melting Pt: 1610 °C (dec.) Density: 3.99 Storage Temperature: Ambient |
MDL Number:
MFCD00014210 CAS Number: 18820-29-6 EINECS: 242-599-3 Merck Index: 14,5740 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...