Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
12783-02
12783-04
CAAA12783-02EA
161.16
CAD
CAAA12783-02
CAAA12783-04
Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate ≥98%
Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate
Purity: 98+%
CAS No.: 50525-27-4
Molecular Formula: C30H24Cl2N6Ru.6H2O
Formula Weight: 748.63 (640.54anhy)
Physical Form: Crystalline
Appearance: Red-orange
Melting Point: 63-66°
MDL No.: MFCD00149670
Formula:
C₃₀H₃₆Cl₂N₆O₆Ru Melting Pt: 63…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149670 CAS Number: 50525-27-4 EINECS: 238-266-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...