Bis(dibenzylideneacetone)palladium

Supplier: Thermo Scientific Chemicals

Synonyms: Bis(dibenzylideneacetone)palladium(0)

12764-01 12764-03 12764-06
CAAA12764-01EA 55.93 CAD
CAAA12764-01 CAAA12764-03 CAAA12764-06
Bis(dibenzylideneacetone)palladium
Bis(dibenzylideneacetone)palladium

Applications: Hydrogenation, isomerization, carbonylation, oxidation, C-C bond formation


CAS No.: 32005-36-0 Molecular Formula: C34H28O2Pd Formula Weight: 575.00 Physical Form: Powder, Packaged under argon Sensitivity: Air and Moisture Sensitive Appearance: Brown to purple Melting Point: 135° dec. Solubility: Slightly soluble in CH2Cl2, CHCl3 and benzene MDL No.: MFCD00051942


Applications: Hydrogenation, isomerization, carbonylation, oxidation, C-C bond formation

Formula: C₃₄H₂₈O₂Pd
MW: 575 g/mol
Melting Pt: 135 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051942
CAS Number: 32005-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR