Dichloro(1,5-cyclooctadiene)platinum(II) (51,6 - 52,6% Pt)
Supplier: Thermo Scientific Chemicals
|
Warning
|
12739-01
12739-03
12739-06
CAAA12739-01EA
96.22
CAD
CAAA12739-01
CAAA12739-03
CAAA12739-06
Dichloro(1,5-cyclooctadiene)platinum(II) (51,6 - 52,6% Pt)
Dichloro(1,5-cyclooctadiene)platinum(II)
Soluble in dichloromethane
CAS No.: 12080-32-9
Molecular Formula: C8H12Cl2Pt
Formula Weight: 374.18
Physical Form: Solid
Sensitivity: Hygroscopic
Melting Point: 250° dec.
Solubility: Soluble in dichloromethane
MDL No.: MFCD00012413
Formula:
C₈H₁₂Cl₂Pt Melting Pt: 250 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012413 CAS Number: 12080-32-9 EINECS: 235-144-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...