Aluminium oxide 20% in water, colloidal dispersion

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

12733-4Z 12733-A1
CAAA12733-4ZEA 295.46 CAD
CAAA12733-4Z CAAA12733-A1
Aluminium oxide 20% in water, colloidal dispersion
Aluminium oxide

Uses: Effective in high temperature refractories as a stable binder
Exists as pseudo-Boehmite with chemical composition corresponding to (AlO(OH) or Al2O3.H2O). Particle charge (+), pH 4, Specific Gravity 1.19, viscosity 10cps, counter ion (mol/mol) 0.049


CAS No.: 1344-28-1 Molecular Formula: Al2O3 Physical Form: 0.05 Micron Particles Odor: Odorless MDL No.: MFCD00003424


Applications: Effective in high temperature refractories as a stable binder

Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980
Melting Pt: 2045
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
Merck Index: 13,00355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR