Potassium tetrabromoaurate(III) dihydrate (≥33% Au) ≥99.99% (metals basis), Premion®

Supplier: Thermo Scientific Chemicals
Warning

12664-03 12664-06
CAAA12664-03EA 170.64 CAD
CAAA12664-03 CAAA12664-06
Potassium tetrabromoaurate(III) dihydrate (≥33% Au) ≥99.99% (metals basis), Premion®
Potassium tetrabromoaurate(III) dihydrate

Soluble in water and alcohol


Purity: 99.99% CAS No.: 14323-32-1 Molecular Formula: KAuBr4.2H2O Formula Weight: 591.72 (555.69anhy) Physical Form: Crystalline Odor: Weak Solubility: Soluble in water and alcohol MDL No.: MFCD00049650

Formula: KAuBr₄.2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00049650
CAS Number: 14323-32-1
EINECS: 238-268-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR