Ruthenium(III) nitrosylnitrate solution (1.5% Ru) (w/v)

Supplier: Thermo Scientific Chemicals
Danger

12630-AD 12630-AK 12630-AU
CAAA12630-ADEA 191.18 CAD
CAAA12630-AD CAAA12630-AK CAAA12630-AU
Ruthenium(III) nitrosylnitrate solution (1.5% Ru) (w/v)
Ruthenium(III) nitrosylnitrate

CAS No.: 34513-98-9 Molecular Formula: N4O10Ru Formula Weight: 317.09 Physical Form: Liquid Density: 1.07 Appearance: Red-brown Odor: Acidic UN No.: 3093 MDL No.: MFCD00016313

Formula: N₄O₁₀Ru
MW: 317.09 g/mol
Density: 1.07
Storage Temperature: Ambient
MDL Number: MFCD00016313
CAS Number: 34513-98-9
EINECS: 252-068-8
UN: 3093
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR